missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-aminobenzoate, 98%

CAS: 94-09-7 | C9H11NO2 | 165.19 g/mol

$89.55 - $310.51

Chemical Identifiers

CAS 94-09-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007892
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES CCOC(=O)C1=CC=C(N)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC150781000
SDS View Documents
Thermo Scientific Chemicals
150781000
100 g Plastic bottle
Each for $89.55
Only null left
 
AC150785000
SDS View Documents
Thermo Scientific Chemicals
150785000
500 g Plastic bottle
Each for $310.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

94-09-7
165.19
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
C9H11NO2
MFCD00007892
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(N)C=C1

Specifications

94-09-7
White
>100°C
97.5% min. (GC)
C9H11NO2
MFCD00007892
14,422
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
165.19
Crystalline Powder
89.0°C to 92.0°C
172°C (17.0 hPa)
Authentic
Plastic bottle
H2NC6H4CO2C2H5
100 g
15,1088
Solubility in water: 1g/2500mL. Other solubilities: 1g/5mL alcohol,1g/2mL chloroform,1g/4mL ether,ca. 1g/40mL expressed almond oil or olive oil
CCOC(=O)C1=CC=C(N)C=C1
165.19
CHEBI:116735
98%
Ethyl 4-aminobenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 202-303-5

SDS
Documents

Documents

RUO – Research Use Only