missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-methyl-4-pentenoate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427370010

 View more versions of this product

Catalog No. AC427370010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl 4-methyl-4-pentenoate
0.8910g/mL
47°C
Glass bottle
CH2=C(CH3)CH2CH2C(O)OCH2CH3
1g
ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd
CCOC(=O)CCC(=C)C
142.2
142.2
4911-54-0
85.0°C (20.0 mmHg)
94% min. (GC)
C8H14O2
MFCD00075160
0.891
RRTBSPBUHUUTHR-UHFFFAOYSA-N
ethyl 4-methylpent-4-enoate
3416433
95%

Chemical Identifiers

4911-54-0
142.2
RRTBSPBUHUUTHR-UHFFFAOYSA-N
3416433
CCOC(=O)CCC(=C)C
C8H14O2
MFCD00075160
ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd
ethyl 4-methylpent-4-enoate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only