missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-methyl-4-pentenoate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 4911-54-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.2
MDL Number MFCD00075160
InChI Key RRTBSPBUHUUTHR-UHFFFAOYSA-N
Synonym ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd
PubChem CID 3416433
IUPAC Name ethyl 4-methylpent-4-enoate
SMILES CCOC(=O)CCC(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427370010
SDS View Documents
Thermo Scientific Chemicals
427370010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427370050
SDS View Documents
Thermo Scientific Chemicals
427370050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4911-54-0
142.2
RRTBSPBUHUUTHR-UHFFFAOYSA-N
3416433
CCOC(=O)CCC(=C)C
C8H14O2
MFCD00075160
ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd
ethyl 4-methylpent-4-enoate

Specifications

4911-54-0
85.0°C (20.0 mmHg)
94% min. (GC)
C8H14O2
MFCD00075160
0.891
RRTBSPBUHUUTHR-UHFFFAOYSA-N
ethyl 4-methylpent-4-enoate
3416433
95%
0.8910g/mL
47°C
Glass bottle
CH2=C(CH3)CH2CH2C(O)OCH2CH3
1g
ethyl 4-methyl-4-pentenoate, 4-methyl-pent-4-enoic acid ethyl ester, 4-pentenoic acid,4-methyl-, ethyl ester, acmc-20akyd
CCOC(=O)CCC(=C)C
142.2
142.2
Ethyl 4-methyl-4-pentenoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only