missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 168421000

 View more versions of this product

Catalog No. AC168421000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 4-nitrobenzoate
99-77-4
Yellow
98.5% min. (GC)
C9H9NO4
MFCD00007351
09, 390
ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate
PHWSCBWNPZDYRI-UHFFFAOYSA-N
ethyl 4-nitrobenzoate
7457
99%
99%
56.0°C to 59.0°C
Authentic
Glass bottle
O2NC6H4CO2C2H5
100 g
10, 3776
Solubility in water: insoluble. Other solubilities: freely soluble in alcohol and ether
CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
195.17
195.17
Crystalline Powder

Chemical Identifiers

99-77-4
195.17
PHWSCBWNPZDYRI-UHFFFAOYSA-N
7457
CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-]
C9H9NO4
MFCD00007351
ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate
ethyl 4-nitrobenzoate
Safety and Handling

Safety and Handling

EINECSNumber : 202-786-2

RTECSNumber : DH5600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only