missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
$137.29 - $778.89
Chemical Identifiers
CAS | 99-77-4 |
---|---|
Molecular Formula | C9H9NO4 |
Molecular Weight (g/mol) | 195.17 |
MDL Number | MFCD00007351 |
InChI Key | PHWSCBWNPZDYRI-UHFFFAOYSA-N |
Synonym | ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate |
PubChem CID | 7457 |
IUPAC Name | ethyl 4-nitrobenzoate |
SMILES | CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC168421000
|
Thermo Scientific Chemicals
168421000 |
100 g | Glass bottle |
N/A
|
|
||||
AC168425000
|
Thermo Scientific Chemicals
168425000 |
500 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-77-4 | |
195.17 | |
PHWSCBWNPZDYRI-UHFFFAOYSA-N | |
7457 | |
CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-] |
C9H9NO4 | |
MFCD00007351 | |
ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate | |
ethyl 4-nitrobenzoate |
Specifications
99-77-4 | |
Yellow | |
98.5% min. (GC) | |
C9H9NO4 | |
MFCD00007351 | |
09, 390 | |
ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate | |
PHWSCBWNPZDYRI-UHFFFAOYSA-N | |
ethyl 4-nitrobenzoate | |
7457 | |
99% | |
Ethyl 4-nitrobenzoate, 99% |
56.0°C to 59.0°C | |
Authentic | |
Glass bottle | |
O2NC6H4CO2C2H5 | |
100 g | |
10, 3776 | |
Solubility in water: insoluble. Other solubilities: freely soluble in alcohol and ether | |
CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-] | |
195.17 | |
195.17 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 202-786-2
RTECSNumber : DH5600000
TSCA : TSCA
RUO – Research Use Only