missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 4-nitrobenzoate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 168425000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 4-nitrobenzoate | |
99-77-4 | |
Yellow | |
98.5% min. (GC) | |
C9H9NO4 | |
MFCD00007351 | |
09, 390 | |
ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate | |
PHWSCBWNPZDYRI-UHFFFAOYSA-N | |
ethyl 4-nitrobenzoate | |
7457 | |
99% |
99% | |
56.0°C to 59.0°C | |
Authentic | |
Glass bottle | |
O2NC6H4CO2C2H5 | |
500 g | |
10, 3776 | |
Solubility in water: insoluble. Other solubilities: freely soluble in alcohol and ether | |
CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-] | |
195.17 | |
195.17 | |
Crystalline Powder |
Chemical Identifiers
99-77-4 | |
195.17 | |
PHWSCBWNPZDYRI-UHFFFAOYSA-N | |
7457 | |
CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-] |
C9H9NO4 | |
MFCD00007351 | |
ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate | |
ethyl 4-nitrobenzoate |
Safety and Handling
EINECSNumber : 202-786-2
RTECSNumber : DH5600000
TSCA : TSCA
RUO – Research Use Only