Learn More


Ethyl Acetate (Certified ACS), Fisher Chemical™
Meet or exceeds the latest ACS specifications. Actual lot of analysis on label
Supplier: Thermo Fisher Scientific BPE1454
Specifications
Ethyl Acetate | |
141-78-6 | |
0.902g/cm³ | |
4 L | |
Glass Bottle | |
0.003% max. | |
0.0009mEq/g max. | |
MFCD00009171 | |
0.45 cP at 20°C | |
0.1% max. | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11 | |
Certified ACS Reagent |
-83.5°C | |
Colorless | |
75°C | |
Pass Test | |
Liquid | |
≥99.5 % | |
C4H8O2 | |
103mbar at 20°C | |
1173 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 | |
≥99.5% |
Chemical Identifiers
141-78-6 | |
88.106 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
8857 | |
ethyl acetate |
C4H8O2 | |
MFCD00009171 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CHEBI:27750 | |
CCOC(=O)C |
Safety and Handling


DANGER!
Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.
NFPA
Health:1
Flammability:3
Instability:0
DANGER!
DOTInformation : DOT Class 3, : Flammable Liquid