missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate (Certified ACS), Fisher Chemical™

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

$541.00 - $19019.73

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
E145-4
SDS Accessories View Documents
Fisher Chemical
BPE1454
4 L Glass Bottle
Each for $541.00
Case of 4 Each for $2,164.00
Only null left
 
E145-20
SDS View Documents
Fisher Chemical
HGL2146
20 L Steel Pail
Each for $1,932.18
Only null left
 
E145-200
SDS Accessories View Documents
Fisher Chemical
E145200
200 L Steel Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C

Specifications

-83.5°C
Colorless
75°C
Pass Test
Liquid
≥99.5 %
C4H8O2
103mbar at 20°C
1173
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CCOC(=O)C
88.106
CHEBI:27750
≥99.5%
Ethyl Acetate
141-78-6
0.902g/cm³
4 L
Glass Bottle
0.003% max.
0.0009mEq/g max.
MFCD00009171
0.45 cP at 20°C
0.1% max.
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
Certified ACS Reagent
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'