missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl chloroformate, 99%, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 430791000

 View more versions of this product

Catalog No. AC430791000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Specifications

Ethyl chloroformate
-81.0°C
1.1350g/mL
10°C
1% max. Diethyl carbonate (GC)
C3H5ClO2
ClCO2C2H5
100 mL
15,3839
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in benzene, chloroform and ether, soluble in most organic solvents
CCOC(=O)Cl
108.52
0.46 mPa.s (20°C)
99%
541-41-3
White
94.0°C
Authentic
AcroSeal™ Glass bottle
1.3950 to 1.3960
MFCD00000644
1.135
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
RIFGWPKJUGCATF-UHFFFAOYSA-N
ethyl carbonochloridate
10928
108.52
Crystalline Solid

Chemical Identifiers

541-41-3
108.52
RIFGWPKJUGCATF-UHFFFAOYSA-N
10928
CCOC(=O)Cl
C3H5ClO2
MFCD00000644
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato
ethyl carbonochloridate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

EINECSNumber : 208-778-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only