Learn More
Ethyl chloroformate, 99%, AcroSeal™, Thermo Scientific Chemicals
$65.31 - $65.31
Chemical Identifiers
CAS | 541-41-3 |
---|---|
Molecular Formula | C3H5ClO2 |
Molecular Weight (g/mol) | 108.52 |
MDL Number | MFCD00000644 |
InChI Key | RIFGWPKJUGCATF-UHFFFAOYSA-N |
Synonym | ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato |
PubChem CID | 10928 |
IUPAC Name | ethyl carbonochloridate |
SMILES | CCOC(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC430791000
|
Thermo Scientific Chemicals
430791000 |
100 mL | AcroSeal™ Glass bottle |
Each for $65.31
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
541-41-3 | |
108.52 | |
RIFGWPKJUGCATF-UHFFFAOYSA-N | |
10928 | |
CCOC(=O)Cl |
C3H5ClO2 | |
MFCD00000644 | |
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato | |
ethyl carbonochloridate |
Specifications
541-41-3 | |
White | |
94.0°C | |
Authentic | |
AcroSeal™ Glass bottle | |
1.3950 to 1.3960 | |
MFCD00000644 | |
1.135 | |
ethyl chloroformate, ethyl chlorocarbonate, cathyl chloride, ethoxycarbonyl chloride, chloroformic acid ethyl ester, ethylchloroformate, carbonochloridic acid, ethyl ester, chloro ethoxy methanone, ethylchloorformiaat, etil cloroformiato | |
RIFGWPKJUGCATF-UHFFFAOYSA-N | |
ethyl carbonochloridate | |
10928 | |
108.52 | |
Crystalline Solid |
-81.0°C | |
1.1350g/mL | |
10°C | |
1% max. Diethyl carbonate (GC) | |
C3H5ClO2 | |
ClCO2C2H5 | |
100 mL | |
15,3839 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in benzene, chloroform and ether, soluble in most organic solvents | |
CCOC(=O)Cl | |
108.52 | |
0.46 mPa.s (20°C) | |
99% | |
Ethyl chloroformate |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi
GHS Signal Word: Danger
EINECSNumber : 208-778-5
RUO – Research Use Only