missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl cinnamate, 98% trans, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150531000

 View more versions of this product

Catalog No. AC150531000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl cinnamate
1mg KOH/g max.
Colorless to Yellow
271.0°C
Authentic
C11H12O2
C6H5CH=CHCO2C2H5
1.046
ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
KBEBGUQPQBELIU-CMDGGOBGSA-N
ethyl (E)-3-phenylprop-2-enoate
637758
176.21
Liquid
103-36-6
6.5°C to 7.5°C
1.0460g/mL
135°C
Glass bottle
1.5580 to 1.5610
100 g
15,2299
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CCOC(=O)C=CC1=CC=CC=C1
176.21
CHEBI:4895
98% trans

Chemical Identifiers

103-36-6
176.21
ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
CHEBI:4895
CCOC(=O)C=CC1=CC=CC=C1
C11H12O2
KBEBGUQPQBELIU-CMDGGOBGSA-N
637758
ethyl (E)-3-phenylprop-2-enoate
Safety and Handling

Safety and Handling

EINECSNumber : 203-104-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only