missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl cinnamate, 98% trans
CAS: 103-36-6 | C11H12O2 | 176.21 g/mol
$56.85 - $898.05
Chemical Identifiers
| CAS | 103-36-6 |
|---|---|
| Molecular Formula | C11H12O2 |
| Molecular Weight (g/mol) | 176.21 |
| InChI Key | KBEBGUQPQBELIU-CMDGGOBGSA-N |
| Synonym | ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate |
| PubChem CID | 637758 |
| ChEBI | CHEBI:4895 |
| IUPAC Name | ethyl (E)-3-phenylprop-2-enoate |
| SMILES | CCOC(=O)C=CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC150531000
|
Thermo Scientific Chemicals
150531000 |
100 g | Glass bottle |
Each for $56.85
|
|
||||
|
AC150535000
|
Thermo Scientific Chemicals
150535000 |
500 g | Glass bottle |
Each for $248.44
|
|
||||
|
AC150530025
|
Thermo Scientific Chemicals
150530025 |
2.5 kg | Glass bottle |
Each for $898.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 103-36-6 | |
| 176.21 | |
| ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate | |
| CHEBI:4895 | |
| CCOC(=O)C=CC1=CC=CC=C1 |
| C11H12O2 | |
| KBEBGUQPQBELIU-CMDGGOBGSA-N | |
| 637758 | |
| ethyl (E)-3-phenylprop-2-enoate |
Specifications
| 103-36-6 | |
| 6.5°C to 7.5°C | |
| 1.0460g/mL | |
| 135°C | |
| Glass bottle | |
| 1.5580 to 1.5610 | |
| 100 g | |
| 15,2299 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
| CCOC(=O)C=CC1=CC=CC=C1 | |
| 176.21 | |
| CHEBI:4895 | |
| 98% trans | |
| Ethyl cinnamate |
| 1mg KOH/g max. | |
| Colorless to Yellow | |
| 271.0°C | |
| Authentic | |
| C11H12O2 | |
| C6H5CH=CHCO2C2H5 | |
| 1.046 | |
| ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate | |
| KBEBGUQPQBELIU-CMDGGOBGSA-N | |
| ethyl (E)-3-phenylprop-2-enoate | |
| 637758 | |
| 176.21 | |
| Liquid |
Safety and Handling
EINECSNumber : 203-104-6
RUO – Research Use Only