missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl cyclopropanecarboxylate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2487022

 View more versions of this product

Catalog No. AAB2487022


Explore available promotions

Description

Description

Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Specifications

Ethyl cyclopropanecarboxylate
0.96
18°C (64°F)
1.42
100 g
2039752
Not miscible in water.
CCOC(=O)C1CC1
114.14
114.15
4606-07-9
133°C to 134°C
C6H10O2
MFCD00001282
UN3272
cyclopropanecarboxylic acid ethyl ester, ethyl cyclopropylcarboxylate, ethyl cyclopropane carboxylate, cyclopropanecarboxylic acid, ethyl ester, pubchem11840, ethyl cylopropylcarboxylate, ethoxycarbonyl cyclopropane, ethylcyclopropane carboxylate, ksc497s5p, lddosdvzpsglfz-uhfffaoysa
LDDOSDVZPSGLFZ-UHFFFAOYSA-N
ethyl cyclopropanecarboxylate
78351
98%

Chemical Identifiers

4606-07-9
MFCD00001282
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H225-H302+H312+H332

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 225-010-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only