Learn More
Ethyl cyclopropanecarboxylate, 98%, Thermo Scientific Chemicals
$90.41 - $831.45
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2487014
|
Thermo Scientific Chemicals
B2487014 |
25 g |
Each for $90.41
|
|
|||||
AAB2487022
|
Thermo Scientific Chemicals
B2487022 |
100 g |
Each for $276.60
|
|
|||||
AAB2487036
|
Thermo Scientific Chemicals
B2487036 |
500 g |
Each for $831.45
|
|
|||||
Description
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Chemical Identifiers
4606-07-9 | |
MFCD00001282 |
Specifications
4606-07-9 | |
133°C to 134°C | |
C6H10O2 | |
MFCD00001282 | |
UN3272 | |
cyclopropanecarboxylic acid ethyl ester, ethyl cyclopropylcarboxylate, ethyl cyclopropane carboxylate, cyclopropanecarboxylic acid, ethyl ester, pubchem11840, ethyl cylopropylcarboxylate, ethoxycarbonyl cyclopropane, ethylcyclopropane carboxylate, ksc497s5p, lddosdvzpsglfz-uhfffaoysa | |
LDDOSDVZPSGLFZ-UHFFFAOYSA-N | |
ethyl cyclopropanecarboxylate | |
78351 | |
98% |
0.96 | |
18°C (64°F) | |
1.42 | |
25 g | |
2039752 | |
Not miscible in water. | |
CCOC(=O)C1CC1 | |
114.14 | |
114.15 | |
Ethyl cyclopropanecarboxylate |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H225-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 225-010-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only