Learn More
Ethyl ethynyl ether, 50 wt% solution in hexanes, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 307390010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl ethynyl ether | |
Orange to Brown | |
55.0°C to 56.0°C | |
Glass bottle | |
C2H5OC≡CH | |
1 g | |
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene | |
CCOC#C | |
70.09 | |
70.09 |
927-80-0,92112-69-1 | |
0.7180g/mL | |
−28°C | |
C4H6O | |
MFCD00009247 | |
0.718 | |
WMYNMYVRWWCRPS-UHFFFAOYSA-N | |
ethynoxyethane | |
61239 | |
Liquid |
Chemical Identifiers
927-80-0 | |
70.09 | |
WMYNMYVRWWCRPS-UHFFFAOYSA-N | |
61239 | |
CCOC#C |
C4H6O | |
MFCD00009247 | |
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene | |
ethynoxyethane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.
Suspected of damaging fertility.
May be fatal if swallowed and enters airways.
May cause dam
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
EINECSNumber : 213-164-5
RTECSNumber : KN9900000
RUO – Research Use Only