missing translation for 'onlineSavingsMsg'
Learn More

Ethyl ethynyl ether, 50 wt% solution in hexanes

CAS: 927-80-0 | C4H6O | 70.09 g/mol

$74.36 - $205.12

Chemical Identifiers

CAS 927-80-0
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00009247
InChI Key WMYNMYVRWWCRPS-UHFFFAOYSA-N
Synonym ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
PubChem CID 61239
IUPAC Name ethynoxyethane
SMILES CCOC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC307390010
SDS View Documents
Thermo Scientific Chemicals
307390010
1 g Glass bottle
Each for $74.36
Only null left
 
AC307390050
SDS View Documents
Thermo Scientific Chemicals
307390050
5 g Glass bottle
Each for $205.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

927-80-0
70.09
WMYNMYVRWWCRPS-UHFFFAOYSA-N
61239
CCOC#C
C4H6O
MFCD00009247
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
ethynoxyethane

Specifications

927-80-0,92112-69-1
0.7180g/mL
−28°C
C4H6O
MFCD00009247
0.718
WMYNMYVRWWCRPS-UHFFFAOYSA-N
ethynoxyethane
61239
Liquid
Orange to Brown
55.0°C to 56.0°C
Glass bottle
C2H5OC≡CH
1 g
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
CCOC#C
70.09
70.09
Ethyl ethynyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.
Suspected of damaging fertility.
May be fatal if swallowed and enters airways.
May cause dam

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 213-164-5

RTECSNumber : KN9900000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only