missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl ethynyl ether, 50 wt% solution in hexanes, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 307390050

 View more versions of this product

Catalog No. AC307390050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl ethynyl ether
Orange to Brown
55.0°C to 56.0°C
Glass bottle
C2H5OC≡CH
5 g
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
CCOC#C
70.09
70.09
927-80-0,92112-69-1
0.7180g/mL
−28°C
C4H6O
MFCD00009247
0.718
WMYNMYVRWWCRPS-UHFFFAOYSA-N
ethynoxyethane
61239
Liquid

Chemical Identifiers

927-80-0
70.09
WMYNMYVRWWCRPS-UHFFFAOYSA-N
61239
CCOC#C
C4H6O
MFCD00009247
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
ethynoxyethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing cancer.
Suspected of damaging fertility.
May be fatal if swallowed and enters airways.
May cause dam

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 213-164-5

RTECSNumber : KN9900000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only