missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Glycidyl Ether 98.0+%, TCI America™

Supplier:  TCI America E046525ML

 View more versions of this product

Catalog No. E046525ML


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Glycidyl Ether
Yellow
C5H10O2
25 mL
ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018
CCOCC1CO1
102.13
102.13
Liquid
4016-11-9
128°C
MFCD00085264
2752
HQCSZRIVJVOYSU-UHFFFAOYNA-N
2-(ethoxymethyl)oxirane
19919
≥98.0% (GC)

Chemical Identifiers

4016-11-9
102.13
HQCSZRIVJVOYSU-UHFFFAOYNA-N
19919
CCOCC1CO1
C5H10O2
MFCD00085264
ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018
2-(ethoxymethyl)oxirane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0391

RTECSNumber : TZ3200000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions