missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Glycidyl Ether 98.0+%, TCI America™
$167.76 - $1316.02
Chemical Identifiers
| CAS | 4016-11-9 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.13 |
| MDL Number | MFCD00085264 |
| InChI Key | HQCSZRIVJVOYSU-UHFFFAOYNA-N |
| Synonym | ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018 |
| PubChem CID | 19919 |
| IUPAC Name | 2-(ethoxymethyl)oxirane |
| SMILES | CCOCC1CO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E046525ML
|
TCI America
E046525ML |
25 mL |
Each for $167.76
|
|
|||||
|
E0465500ML
|
TCI America
E0465500ML |
500 mL |
Each for $1,316.02
|
|
|||||
Chemical Identifiers
| 4016-11-9 | |
| 102.13 | |
| HQCSZRIVJVOYSU-UHFFFAOYNA-N | |
| 19919 | |
| CCOCC1CO1 |
| C5H10O2 | |
| MFCD00085264 | |
| ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018 | |
| 2-(ethoxymethyl)oxirane |
Specifications
| 4016-11-9 | |
| 128°C | |
| MFCD00085264 | |
| 2752 | |
| HQCSZRIVJVOYSU-UHFFFAOYNA-N | |
| 2-(ethoxymethyl)oxirane | |
| 19919 | |
| ≥98.0% (GC) | |
| Ethyl Glycidyl Ether |
| Yellow | |
| C5H10O2 | |
| 25 mL | |
| ethyl glycidyl ether, 2-ethoxymethyl oxirane, oxirane, ethoxymethyl, 1,2-epoxy-3-ethoxypropane, ethoxymethyl oxirane, 3-ethoxy-1,2-epoxypropane, oxirane, 2-ethoxymethyl, propane, 1,2-epoxy-3-ethoxy, 1-ethoxy-2,3-epoxypropane, ccris 1018 | |
| CCOCC1CO1 | |
| 102.13 | |
| 102.13 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0391
RTECSNumber : TZ3200000
TSCA : Yes