missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl heptanoate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1472936

 View more versions of this product

Catalog No. AAA1472936


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl heptanoate
-66°C
188°C to 189°C
C9H18O2
MFCD00009538
1752311
ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate
CCCCCCC(=O)OCC
158.24
CHEBI:86618
≥98%
106-30-9
0.868
66°C (151°F)
1.412
500 g
14,3835
TVQGDYNRXLTQAP-UHFFFAOYSA-N
ethyl heptanoate
7797
158.24

Chemical Identifiers

106-30-9
158.24
TVQGDYNRXLTQAP-UHFFFAOYSA-N
7797
ethyl heptanoate
C9H18O2
MFCD00009538
ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate
CHEBI:86618
CCCCCCC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 203-382-9

RTECSNumber : MJ2087000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only