missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl heptanoate, 98+%, Thermo Scientific Chemicals
$59.18 - $189.92
Chemical Identifiers
CAS | 106-30-9 |
---|---|
Molecular Formula | C9H18O2 |
Molecular Weight (g/mol) | 158.24 |
MDL Number | MFCD00009538 |
InChI Key | TVQGDYNRXLTQAP-UHFFFAOYSA-N |
Synonym | ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate |
PubChem CID | 7797 |
ChEBI | CHEBI:86618 |
IUPAC Name | ethyl heptanoate |
SMILES | CCCCCCC(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1472922
|
Thermo Scientific Chemicals
A1472922 |
100 g |
Each for $59.18
|
|
||||||
AAA1472936
|
Thermo Scientific Chemicals
A1472936 |
500 g |
Each for $189.92
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-30-9 | |
158.24 | |
TVQGDYNRXLTQAP-UHFFFAOYSA-N | |
7797 | |
ethyl heptanoate |
C9H18O2 | |
MFCD00009538 | |
ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate | |
CHEBI:86618 | |
CCCCCCC(=O)OCC |
Specifications
106-30-9 | |
0.868 | |
66°C (151°F) | |
1.412 | |
100 g | |
14,3835 | |
TVQGDYNRXLTQAP-UHFFFAOYSA-N | |
ethyl heptanoate | |
7797 | |
158.24 | |
Ethyl heptanoate |
-66°C | |
188°C to 189°C | |
C9H18O2 | |
MFCD00009538 | |
1752311 | |
ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate | |
CCCCCCC(=O)OCC | |
158.24 | |
CHEBI:86618 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 203-382-9
RTECSNumber : MJ2087000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only