missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isobutyrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 310515000

 View more versions of this product

Catalog No. AC310515000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl isobutyrate
97-62-1
0.8600g/mL
13°C
99%
C6H12O2
(CH3)2CHCO2C2H5
500 mL
15,3869
Solubility in water: not miscible
CCOC(=O)C(C)C
116.18
CHEBI:87303
99%
99%
Colorless
112.0°C to 113.0°C
Authentic
Glass bottle
1.3860 to 1.3880
MFCD00009165
0.86
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
WDAXFOBOLVPGLV-UHFFFAOYSA-N
ethyl 2-methylpropanoate
7342
116.18
Liquid

Chemical Identifiers

97-62-1
116.18
WDAXFOBOLVPGLV-UHFFFAOYSA-N
7342
ethyl 2-methylpropanoate
C6H12O2
MFCD00009165
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
CHEBI:87303
CCOC(=O)C(C)C
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Danger

EINECSNumber : 202-595-4

RTECSNumber : NQ4675000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only