missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isobutyrate, 99%, Thermo Scientific Chemicals

$78.25 - $78.25

Chemical Identifiers

CAS 97-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.18
MDL Number MFCD00009165
InChI Key WDAXFOBOLVPGLV-UHFFFAOYSA-N
Synonym ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
PubChem CID 7342
ChEBI CHEBI:87303
IUPAC Name ethyl 2-methylpropanoate
SMILES CCOC(=O)C(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC310515000
SDS View Documents
Thermo Scientific Chemicals
310515000
500 mL Glass bottle
Each for $78.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97-62-1
116.18
WDAXFOBOLVPGLV-UHFFFAOYSA-N
7342
ethyl 2-methylpropanoate
C6H12O2
MFCD00009165
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
CHEBI:87303
CCOC(=O)C(C)C

Specifications

97-62-1
0.8600g/mL
13°C
99%
C6H12O2
(CH3)2CHCO2C2H5
500 mL
15,3869
Solubility in water: not miscible
CCOC(=O)C(C)C
116.18
CHEBI:87303
99%
Ethyl isobutyrate, 99%
Colorless
112.0°C to 113.0°C
Authentic
Glass bottle
1.3860 to 1.3880
MFCD00009165
0.86
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
WDAXFOBOLVPGLV-UHFFFAOYSA-N
ethyl 2-methylpropanoate
7342
116.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Danger

EINECSNumber : 202-595-4

RTECSNumber : NQ4675000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only