missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl isobutyrylacetate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 383440500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl isobutyrylacetate | |
-9.0°C | |
0.9810g/mL | |
44°C | |
94% min. (Mixture of tautomers) (GC) | |
C8H14O3 | |
(CH3)2CHCOCH2CO2C2H5 | |
50 g | |
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate | |
CCOC(=O)CC(=O)C(C)C | |
158.2 | |
158.2 | |
Liquid |
7152-15-0 | |
Colorless to Yellow | |
173.0°C | |
Authentic | |
Glass bottle | |
1.4255 to 1.4275 | |
MFCD00009198 | |
0.981 | |
XCLDSQRVMMXWMS-UHFFFAOYSA-N | |
ethyl 4-methyl-3-oxopentanoate | |
81583 | |
95% |
Chemical Identifiers
7152-15-0 | |
158.2 | |
XCLDSQRVMMXWMS-UHFFFAOYSA-N | |
81583 | |
CCOC(=O)CC(=O)C(C)C |
C8H14O3 | |
MFCD00009198 | |
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate | |
ethyl 4-methyl-3-oxopentanoate |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 230-491-9
RUO – Research Use Only