missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Levulinate 98.0+%, TCI America™

Supplier:  TCI America L012025ML

 View more versions of this product

Catalog No. L012025ML


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Levulinate
Yellow
C7H12O3
25 mL
GMEONFUTDYJSNV-UHFFFAOYSA-N
ethyl 4-oxopentanoate
10883
≥98.0% (GC)
539-88-8
205°C
MFCD00009209
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
CCOC(=O)CCC(C)=O
144.17
144.17
Liquid

Chemical Identifiers

539-88-8
144.17
GMEONFUTDYJSNV-UHFFFAOYSA-N
10883
CCOC(=O)CCC(C)=O
C7H12O3
MFCD00009209
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
ethyl 4-oxopentanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1492&(2)-1505

RTECSNumber : OI1700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions