missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Levulinate 98.0+%, TCI America™
$41.02 - $177.08
Chemical Identifiers
| CAS | 539-88-8 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00009209 |
| InChI Key | GMEONFUTDYJSNV-UHFFFAOYSA-N |
| Synonym | ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate |
| PubChem CID | 10883 |
| IUPAC Name | ethyl 4-oxopentanoate |
| SMILES | CCOC(=O)CCC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
L012025ML
|
TCI America
L012025ML |
25 mL |
Each for $41.02
|
|
|||||
|
L0120100ML
|
TCI America
L0120100ML |
100 mL |
Each for $67.11
|
|
|||||
|
L0120500ML
|
TCI America
L0120500ML |
500 mL |
Each for $177.08
|
|
|||||
Chemical Identifiers
| 539-88-8 | |
| 144.17 | |
| GMEONFUTDYJSNV-UHFFFAOYSA-N | |
| 10883 | |
| CCOC(=O)CCC(C)=O |
| C7H12O3 | |
| MFCD00009209 | |
| ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate | |
| ethyl 4-oxopentanoate |
Specifications
| 539-88-8 | |
| 205°C | |
| MFCD00009209 | |
| ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate | |
| CCOC(=O)CCC(C)=O | |
| 144.17 | |
| 144.17 | |
| Liquid |
| Yellow | |
| C7H12O3 | |
| 25 mL | |
| GMEONFUTDYJSNV-UHFFFAOYSA-N | |
| ethyl 4-oxopentanoate | |
| 10883 | |
| ≥98.0% (GC) | |
| Ethyl Levulinate |
Safety and Handling
EINECSNumber : (2)-1492&(2)-1505
RTECSNumber : OI1700000
TSCA : Yes