missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Levulinate 98.0+%, TCI America™
Supplier: TCI America L0120500ML
Specifications
Ethyl Levulinate | |
Yellow | |
C7H12O3 | |
500 mL | |
GMEONFUTDYJSNV-UHFFFAOYSA-N | |
ethyl 4-oxopentanoate | |
10883 | |
≥98.0% (GC) |
539-88-8 | |
205°C | |
MFCD00009209 | |
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate | |
CCOC(=O)CCC(C)=O | |
144.17 | |
144.17 | |
Liquid |
Chemical Identifiers
539-88-8 | |
144.17 | |
GMEONFUTDYJSNV-UHFFFAOYSA-N | |
10883 | |
CCOC(=O)CCC(C)=O |
C7H12O3 | |
MFCD00009209 | |
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate | |
ethyl 4-oxopentanoate |
Safety and Handling
EINECSNumber : (2)-1492&(2)-1505
RTECSNumber : OI1700000
TSCA : Yes