missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl oxamate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 118790250

 View more versions of this product

Catalog No. AC118790250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl oxamate
112°C to 115°C
Authentic
Glass bottle
H2NCOCO2C2H5
25 g
ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide
RZMZBHSKPLVQCP-UHFFFAOYSA-N
ethyl 2-amino-2-oxoacetate
69238
99%
617-36-7
White
98.5% min
C4H7NO3
MFCD00008005
02, 544
Solubility in water: soluble
CCOC(=O)C(=O)N
117.1
117.1
Crystalline Powder, Crystals or Chunks

Chemical Identifiers

617-36-7
RZMZBHSKPLVQCP-UHFFFAOYSA-N
69238
CCOC(=O)C(=O)N
MFCD00008005
ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide
ethyl 2-amino-2-oxoacetate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-512-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only