missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Pyrazole-3-carboxylate 98.0+%, TCI America™

Supplier:  TCI America E09655G

 View more versions of this product

Catalog No. E09655G


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Pyrazole-3-carboxylate
161°C
MFCD00159643
Pyrazole-3-carboxylic Acid Ethyl Ester
CCOC(=O)C1=CC=NN1
140.142
140.14
Crystalline Powder
5932-27-4
C6H8N2O2
5 g
MSPOSRHJXMILNK-UHFFFAOYSA-N
ethyl 1H-pyrazole-5-carboxylate
7147518
≥98.0% (GC)

Chemical Identifiers

5932-27-4
140.142
MSPOSRHJXMILNK-UHFFFAOYSA-N
7147518
CCOC(=O)C1=CC=NN1
C6H8N2O2
MFCD00159643
Pyrazole-3-carboxylic Acid Ethyl Ester
ethyl 1H-pyrazole-5-carboxylate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions