missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Pyrazole-3-carboxylate 98.0+%, TCI America™
$39.51 - $116.52
Chemical Identifiers
CAS | 5932-27-4 |
---|---|
Molecular Formula | C6H8N2O2 |
Molecular Weight (g/mol) | 140.142 |
MDL Number | MFCD00159643 |
InChI Key | MSPOSRHJXMILNK-UHFFFAOYSA-N |
Synonym | Pyrazole-3-carboxylic Acid Ethyl Ester |
PubChem CID | 7147518 |
IUPAC Name | ethyl 1H-pyrazole-5-carboxylate |
SMILES | CCOC(=O)C1=CC=NN1 |
Chemical Identifiers
5932-27-4 | |
140.142 | |
MSPOSRHJXMILNK-UHFFFAOYSA-N | |
7147518 | |
CCOC(=O)C1=CC=NN1 |
C6H8N2O2 | |
MFCD00159643 | |
Pyrazole-3-carboxylic Acid Ethyl Ester | |
ethyl 1H-pyrazole-5-carboxylate |
Specifications
5932-27-4 | |
C6H8N2O2 | |
1 g | |
MSPOSRHJXMILNK-UHFFFAOYSA-N | |
ethyl 1H-pyrazole-5-carboxylate | |
7147518 | |
≥98.0% (GC) | |
Ethyl Pyrazole-3-carboxylate |
161°C | |
MFCD00159643 | |
Pyrazole-3-carboxylic Acid Ethyl Ester | |
CCOC(=O)C1=CC=NN1 | |
140.142 | |
140.14 | |
Crystalline Powder |
Safety and Handling
TSCA : No