missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl trans-beta-methylcinnamate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 428000010

 View more versions of this product

Catalog No. AC428000010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl trans-β-methylcinnamate
1.0420g/mL
>113°C
Glass bottle
C6H5C(CH3)=CHCO2C2H5
1 g
e-ethyl 3-phenylbut-2-enoate, cinnamic acid, .beta.-methyl-, ethyl ester, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl trans-beta-methylcinnamate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenyl-2-butenoate, ethyl e-3-phenyl-2-butenoate, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, 2-butenoic acid, 3-phenyl-, ethyl ester
CCOC(=O)C=C(C)C1=CC=CC=C1
190.24
190.24
1504-72-9
142°C to 144°C (20.0 hPa)
96% min. (GC)
C12H14O2
MFCD00053762
1.042
BSXHSWOMMFBMLL-MDZDMXLPSA-N
ethyl (E)-3-phenylbut-2-enoate
5354836
97%

Chemical Identifiers

1504-72-9
190.24
BSXHSWOMMFBMLL-MDZDMXLPSA-N
5354836
CCOC(=O)C=C(C)C1=CC=CC=C1
C12H14O2
MFCD00053762
e-ethyl 3-phenylbut-2-enoate, cinnamic acid, .beta.-methyl-, ethyl ester, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl trans-beta-methylcinnamate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenyl-2-butenoate, ethyl e-3-phenyl-2-butenoate, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, 2-butenoic acid, 3-phenyl-, ethyl ester
ethyl (E)-3-phenylbut-2-enoate
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only