missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl trans-beta-methylcinnamate, 97%, Thermo Scientific Chemicals

$63.81 - $63.81

Chemical Identifiers

CAS 1504-72-9
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00053762
InChI Key BSXHSWOMMFBMLL-MDZDMXLPSA-N
Synonym e-ethyl 3-phenylbut-2-enoate, cinnamic acid, .beta.-methyl-, ethyl ester, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl trans-beta-methylcinnamate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenyl-2-butenoate, ethyl e-3-phenyl-2-butenoate, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, 2-butenoic acid, 3-phenyl-, ethyl ester
PubChem CID 5354836
IUPAC Name ethyl (E)-3-phenylbut-2-enoate
SMILES CCOC(=O)C=C(C)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428000010
SDS View Documents
Thermo Scientific Chemicals
428000010
1 g Glass bottle
Each for $63.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1504-72-9
190.24
BSXHSWOMMFBMLL-MDZDMXLPSA-N
5354836
CCOC(=O)C=C(C)C1=CC=CC=C1
C12H14O2
MFCD00053762
e-ethyl 3-phenylbut-2-enoate, cinnamic acid, .beta.-methyl-, ethyl ester, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl trans-beta-methylcinnamate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenyl-2-butenoate, ethyl e-3-phenyl-2-butenoate, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, 2-butenoic acid, 3-phenyl-, ethyl ester
ethyl (E)-3-phenylbut-2-enoate

Specifications

1504-72-9
142°C to 144°C (20.0 hPa)
96% min. (GC)
C12H14O2
MFCD00053762
1.042
BSXHSWOMMFBMLL-MDZDMXLPSA-N
ethyl (E)-3-phenylbut-2-enoate
5354836
97%
1.0420g/mL
>113°C
Glass bottle
C6H5C(CH3)=CHCO2C2H5
1 g
e-ethyl 3-phenylbut-2-enoate, cinnamic acid, .beta.-methyl-, ethyl ester, ethyl 3-phenylbut-2-enoate, ethyl 3-phenylcrotonate, ethyl trans-beta-methylcinnamate, ethyl trans-beta-methylcinnamate, ethyl e-3-phenyl-2-butenoate, ethyl e-3-phenyl-2-butenoate, a-methylcinnamate, 2-butenoic acid, 3-phenyl-, ethyl ester, 2-butenoic acid, 3-phenyl-, ethyl ester
CCOC(=O)C=C(C)C1=CC=CC=C1
190.24
190.24
Ethyl trans-β-methylcinnamate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only