missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 185451000

 View more versions of this product

Catalog No. AC185451000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Ethylaluminium dichloride
Colorless
AcroSeal™ Glass bottle
C2H5AlCl2
100 mL
0.74
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
UAIZDWNSWGTKFZ-UHFFFAOYSA-L
dichloro(ethyl)alumane
126.95
Liquid
563-43-9,142-82-5
0.7400g/mL
C2H5AlCl2
MFCD00000457
06,251; 07,146; 10,177
15, 3816
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
CC[Al](Cl)Cl
126.94
14.5 to 15.5%

Chemical Identifiers

563-43-9
126.94
UAIZDWNSWGTKFZ-UHFFFAOYSA-L
dichloro(ethyl)alumane
C2H5AlCl2
MFCD00000457
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
CC[Al](Cl)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effec

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 209-248-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only