missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle2-amino-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific™

$41.86 - $41.86

Chemical Identifiers

CAS 81633-29-6
Molecular Formula C8H11N3O2
Molecular Weight (g/mol) 181.20
MDL Number MFCD00052622
InChI Key YBFVMJRSZCVJJP-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyrimidine-5-carboxylic acid ethyl ester, 2-amino-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, acmc-209plx, maybridge1_003176, ethyl2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-5-ethoxycarbonyl-4-methylpyrimidine, ethyl 2-amino-4-methyl-pyrimidine-5-carboxylate, ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-4-methyl-5-pyrimidinecarboxylic acid, ethyl ester, 5-pyrimidinecarboxylicacid, 2-amino-4-methyl-, ethyl ester
PubChem CID 459813
IUPAC Name ethyl 2-amino-4-methylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N)N=C1C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CD10070DA
SDS View Documents
Thermo Scientific Chemicals
CD10070DA
1 g Amber Glass Bottle
Each for $41.86
 
Description

Description

Specifications

Chemical Identifiers

81633-29-6
181.20
YBFVMJRSZCVJJP-UHFFFAOYSA-N
459813
CCOC(=O)C1=CN=C(N)N=C1C
C8H11N3O2
MFCD00052622
2-amino-4-methylpyrimidine-5-carboxylic acid ethyl ester, 2-amino-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, acmc-209plx, maybridge1_003176, ethyl2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-5-ethoxycarbonyl-4-methylpyrimidine, ethyl 2-amino-4-methyl-pyrimidine-5-carboxylate, ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-4-methyl-5-pyrimidinecarboxylic acid, ethyl ester, 5-pyrimidinecarboxylicacid, 2-amino-4-methyl-, ethyl ester
ethyl 2-amino-4-methylpyrimidine-5-carboxylate

Specifications

81633-29-6
100.0
Amber Glass Bottle
MFCD00052622
2-amino-4-methylpyrimidine-5-carboxylic acid ethyl ester, 2-amino-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, acmc-209plx, maybridge1_003176, ethyl2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-5-ethoxycarbonyl-4-methylpyrimidine, ethyl 2-amino-4-methyl-pyrimidine-5-carboxylate, ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-4-methyl-5-pyrimidinecarboxylic acid, ethyl ester, 5-pyrimidinecarboxylicacid, 2-amino-4-methyl-, ethyl ester
CCOC(=O)C1=CN=C(N)N=C1C
181.20
181.19
Ethyl 2-amino-4-methylpyrimidine-5-carboxylate
96.5
97%
C8H11N3O2
1 g
YBFVMJRSZCVJJP-UHFFFAOYSA-N
ethyl 2-amino-4-methylpyrimidine-5-carboxylate
459813
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'