missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene glycol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A115910C

 View more versions of this product

Catalog No. AAA115910C


Explore available promotions

Description

Description

Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.

Solubility
Miscible with water, toluene, tetrahydrofuran, alcohol, chloroform and other organic solvents.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents, strong bases, aldehydes and aluminum.
Specifications

Specifications

Ethylene glycol
107-21-1
196°C to 198°C
119°C (246°F)
1.431
HOCH2CH2OH
505945
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.07
-13°C
1.113
10,000 g
Sweet
C2H6O2
MFCD00002885
14,3798
Miscible with water,toluene,tetrahydrofuran,alcohol,chloroform and other organic solvents.
C(CO)O
62.068
CHEBI:30742
99%

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
MFCD00002885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c

H302-H336-H373

EINECSNumber : 203-473-3

RTECSNumber : KW2975000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only