Learn More
Ethylene glycol, 99%, Thermo Scientific Chemicals
$47.45 - $577.89
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.068 |
MDL Number | MFCD00002885 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
IUPAC Name | ethane-1,2-diol |
SMILES | C(CO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1159130
|
Thermo Scientific Chemicals
A1159130 |
250 g |
Each for $47.45
|
|
|||||
AAA1159136
|
Thermo Scientific Chemicals
A1159136 |
500 g |
Each for $75.32
|
|
|||||
AAA115910E
|
Thermo Scientific Chemicals
A115910E |
2500 g |
Each for $197.62
|
|
|||||
AAA115910C
|
Thermo Scientific Chemicals
A115910C |
10,000 g |
Each for $577.89
|
|
|||||
Description
Ethylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylene glycol is used as a precursor for the syntheses of polymers like polyester fibers, resins and polyethylene terephthalate, which finds application in making plastic bottles. It acts as an intermediate in the synthesis of 1,4-dioxane. It plays an important role in organic synthesis as a protecting group for carbonyl groups. It is a useful desiccant due to its high boiling point and affinity towards water. It plays an important role to remove water vapor from natural gas before proceeding for further processes. Its major application is as a medium for convective heat transfer in automobiles and liquid cooled computers.
Solubility
Miscible with water, toluene, tetrahydrofuran, alcohol, chloroform and other organic solvents.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents, strong bases, aldehydes and aluminum.
Chemical Identifiers
107-21-1 | |
62.068 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
C2H6O2 | |
MFCD00002885 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
Specifications
-13°C | |
1.113 | |
250 g | |
Sweet | |
C2H6O2 | |
MFCD00002885 | |
14,3798 | |
Miscible with water,toluene,tetrahydrofuran,alcohol,chloroform and other organic solvents. | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
99% |
107-21-1 | |
196°C to 198°C | |
119°C (246°F) | |
1.431 | |
HOCH2CH2OH | |
505945 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.07 | |
Ethylene glycol |
Safety and Handling
GHS H Statement
H373-H302
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
H302-H336-H373
EINECSNumber : 203-473-3
RTECSNumber : KW2975000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only