Learn More
Ethylene glycol diethyl ether, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 208240010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethylene glycol diethyl ether | |
-74.0°C | |
0.8417g/mL | |
19°C | |
98% min. (GC) | |
C6H14O2 | |
C2H5OCH2CH2OC2H5 | |
1 L | |
0.8417 | |
Solubility in water: 20.4%. | |
CCOCCOCC | |
118.18 | |
0.7 mPa.s (20°C) | |
99% |
629-14-1 | |
Colorless | |
121.0°C | |
Authentic | |
Glass bottle | |
1.3910 to 1.3930 | |
MFCD00009253 | |
01, 468 | |
ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu | |
LZDKZFUFMNSQCJ-UHFFFAOYSA-N | |
1,2-diethoxyethane | |
12375 | |
118.18 | |
Liquid |
Chemical Identifiers
629-14-1 | |
118.18 | |
LZDKZFUFMNSQCJ-UHFFFAOYSA-N | |
12375 | |
CCOCCOCC |
C6H14O2 | |
MFCD00009253 | |
ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu | |
1,2-diethoxyethane |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May damage the unborn child.
Suspected of damaging fertility.
Highly flammable liquid and vapor.
May form explosive peroxides.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES
GHS Signal Word: Danger
EINECSNumber : 211-076-1
RUO – Research Use Only