missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene Glycol Monoallyl Ether 98.0+%, TCI America™

Supplier:  TCI America E032625ML

 View more versions of this product

Catalog No. E032625ML


Explore available promotions

Description

Description

Specifications

Specifications

Ethylene Glycol Monoallyl Ether
Colorless
C5H10O2
25 mL
2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether
OCCOCC=C
102.13
102.13
Liquid
111-45-5
161°C
MFCD00020607
3271
GCYHRYNSUGLLMA-UHFFFAOYSA-N
2-(prop-2-en-1-yloxy)ethan-1-ol
8116
≥98.0% (GC)

Chemical Identifiers

111-45-5
102.13
GCYHRYNSUGLLMA-UHFFFAOYSA-N
8116
OCCOCC=C
C5H10O2
MFCD00020607
2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether
2-(prop-2-en-1-yloxy)ethan-1-ol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0097&(9)-0123

RTECSNumber : KJ5425000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions