missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Monoallyl Ether 98.0+%, TCI America™
$46.60 - $104.38
Chemical Identifiers
| CAS | 111-45-5 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.13 |
| MDL Number | MFCD00020607 |
| InChI Key | GCYHRYNSUGLLMA-UHFFFAOYSA-N |
| Synonym | 2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether |
| PubChem CID | 8116 |
| IUPAC Name | 2-(prop-2-en-1-yloxy)ethan-1-ol |
| SMILES | OCCOCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E032625ML
|
TCI America
E032625ML |
25 mL |
Each for $46.60
|
|
|||||
|
E0326500ML
|
TCI America
E0326500ML |
500 mL |
Each for $104.38
|
|
|||||
Chemical Identifiers
| 111-45-5 | |
| 102.13 | |
| GCYHRYNSUGLLMA-UHFFFAOYSA-N | |
| 8116 | |
| OCCOCC=C |
| C5H10O2 | |
| MFCD00020607 | |
| 2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether | |
| 2-(prop-2-en-1-yloxy)ethan-1-ol |
Specifications
| 111-45-5 | |
| 161°C | |
| MFCD00020607 | |
| 3271 | |
| GCYHRYNSUGLLMA-UHFFFAOYSA-N | |
| 2-(prop-2-en-1-yloxy)ethan-1-ol | |
| 8116 | |
| ≥98.0% (GC) | |
| Ethylene Glycol Monoallyl Ether |
| Colorless | |
| C5H10O2 | |
| 25 mL | |
| 2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether | |
| OCCOCC=C | |
| 102.13 | |
| 102.13 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-0097&(9)-0123
RTECSNumber : KJ5425000
TSCA : Yes