missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Tetraacetic Acid (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific E478500

 View more versions of this product

Catalog No. E478-500



Explore available promotions

Description

Description

Specifications

Specifications

Ethylenediamine Tetraacetic Acid
220°C
0.86g/mL
0.2% max.
Glass Bottle
(HOOCCH2)2NCH2CH2N(CH2COOH)2
500 g
0.005% max. Insoluble in NH4OH
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
292.24
CHEBI:42191
0.013 hPa at 20°C
Certified ACS
60-00-4
White
2.5
99.4 to 100.6%
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
6049
292.25
99.4 to 100.6%
Powder or Solid

Chemical Identifiers

60-00-4
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions