missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine Tetraacetic Acid (Certified ACS), Fisher Chemical™

$388.75 - $686.83

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
E478-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
E478500
500 g Glass Bottle
Each for $388.75
Case of 6 Each for $2,332.50
 
E478-1
SDS View Documents
Thermo Fisher Scientific
E4781
1 kg Glass Bottle
Each for $686.83
Case of 4 Each for $2,747.32
 
Description

Description

Specifications

Chemical Identifiers

60-00-4
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

60-00-4
White
2.5
99.4 to 100.6%
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
6049
292.25
99.4 to 100.6%
Powder or Solid
220°C
0.86g/mL
0.2% max.
Glass Bottle
(HOOCCH2)2NCH2CH2N(CH2COOH)2
500 g
0.005% max. Insoluble in NH4OH
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
292.24
CHEBI:42191
0.013 hPa at 20°C
Certified ACS
Ethylenediamine Tetraacetic Acid
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'