missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyltriphenylphosphonium Iodide 98.0+%, TCI America™

Supplier:  TCI America E054925G

 View more versions of this product

Catalog No. E054925G


Explore available promotions

Description

Description

Specifications

Specifications

Ethyltriphenylphosphonium Iodide
170°C
C20H20IP
25 g
ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide
[I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
418.26
418.26
Crystalline Powder
4736-60-1
White
MFCD00040352
3464
SLAFUPJSGFVWPP-UHFFFAOYSA-M
ethyltriphenylphosphanium iodide
78474
≥98.0% (HPLC,T)

Chemical Identifiers

4736-60-1
418.26
SLAFUPJSGFVWPP-UHFFFAOYSA-M
78474
[I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C20H20IP
MFCD00040352
ethyltriphenylphosphonium iodide, phosphonium, ethyltriphenyl-, iodide, ethyl triphenylphosphonium iodide, phenylphosphonium ethyl iodide, unii-xxz2v239jn, xxz2v239jn, ethyltriphenylphosphanium iodide, phosphonium, ethyltriphenyl-, iodide 1:1, ethyl triphenyl phosphanium iodide, ethyl triphenyl phosphonium iodide
ethyltriphenylphosphanium iodide
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TA2312000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions