missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethynylcyclopropane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 369130100

 View more versions of this product

Catalog No. AC369130100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethynylcyclopropane
6746-94-7
0.7801g/mL
−17°C
97.5% min. (GC)
C5H6
MFCD02181090
0.7801
Solubility in water: 3.96g/L (20°C). Other solubilities: soluble in ethanol, dimethylsulfoxide, toluene,, acetone
C#CC1CC1
66.1
66.1
Liquid
98%
Colorless to Yellow
51.0°C to 52.0°C
Authentic
Glass bottle
1.4280 to 1.5300
10 g
cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
NPTDXPDGUHAFKC-UHFFFAOYSA-N
ethynylcyclopropane
138823
98%

Chemical Identifiers

6746-94-7
66.1
NPTDXPDGUHAFKC-UHFFFAOYSA-N
138823
C#CC1CC1
C5H6
MFCD02181090
cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
ethynylcyclopropane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only