Learn More
Ethynylcyclopropane, 98%, Thermo Scientific Chemicals
$38.33 - $87.15
Chemical Identifiers
CAS | 6746-94-7 |
---|---|
Molecular Formula | C5H6 |
Molecular Weight (g/mol) | 66.1 |
MDL Number | MFCD02181090 |
InChI Key | NPTDXPDGUHAFKC-UHFFFAOYSA-N |
Synonym | cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene |
PubChem CID | 138823 |
IUPAC Name | ethynylcyclopropane |
SMILES | C#CC1CC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC369130010
|
Thermo Scientific Chemicals
369130010 |
1 g | Glass bottle |
N/A
|
|
||||
AC369130100
|
Thermo Scientific Chemicals
369130100 |
10 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6746-94-7 | |
66.1 | |
NPTDXPDGUHAFKC-UHFFFAOYSA-N | |
138823 | |
C#CC1CC1 |
C5H6 | |
MFCD02181090 | |
cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene | |
ethynylcyclopropane |
Specifications
6746-94-7 | |
0.7801g/mL | |
−17°C | |
97.5% min. (GC) | |
C5H6 | |
MFCD02181090 | |
0.7801 | |
Solubility in water: 3.96g/L (20°C). Other solubilities: soluble in ethanol, dimethylsulfoxide, toluene,, acetone | |
C#CC1CC1 | |
66.1 | |
66.1 | |
Liquid |
Colorless to Yellow | |
51.0°C to 52.0°C | |
Authentic | |
Glass bottle | |
1.4280 to 1.5300 | |
1 g | |
cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene | |
NPTDXPDGUHAFKC-UHFFFAOYSA-N | |
ethynylcyclopropane | |
138823 | |
98% | |
Ethynylcyclopropane, 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only