Bullard™ EVA PAPR System with Double Bib Tychem QC Hoods
Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) use a battery-powered fan to draw in ambient air through a filter cartridge then delivers filtered air to the user's double bib hood via a breathing tube.
Supplier:  Bullard™ EVA20TIC
 View more versions of this product
Catalog No. 17-986-264
Explore available promotions
Description
Description
  • Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
  • Easy-to-read fuel gauge displays remaining battery capacity at a glance
  • Battery status can be checked at any time during use
  • Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
  • Two distinct audible alarms notify wearers of low battery and low air flow
  • Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
  • Powered by a long-lasting 10,000-hour brushless motor
  • Single filter design for easy use
  • Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
  • Filters are sold separately
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: 1.) hood or mask; 2.) breathing tube; 3.) blower assembly; 4.) purifying filter cartridge (sold separately). Check model system components carefully before ordering.
Compliance
NIOSH approved.
Specifications
Specifications
7 to 8 hours
Compatible with PAPRFC3, PAPRFC4, PAPRFC5. (Sold separately.)
EVA blower, battery, belt, charger, breathing tube, Tychem QC Hood
EVA20TIC
Foam
Double bib hood, ratchet suspension
Acetate