Bullard™ EVA PAPR System with Double Bib Tychem QC Hoods
Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) use a battery-powered fan to draw in ambient air through a filter cartridge then delivers filtered air to the user's double bib hood via a breathing tube.
$2490.85 - $2495.96
Catalog Number | Mfr. No. | Facepiece/Headtop Model | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Facepiece/Headtop Model | Price | Quantity | |||||
17-986-264
|
Bullard™
EVA20TIC |
Double bib hood, ratchet suspension |
Each for $2,494.25
|
|
|||||
17-989-615
|
Bullard™
EVART3T |
Double bib hood, for use without headband, quick-attach |
Each for $2,492.54
|
|
|||||
17-986-270
|
Bullard™
EVART3 |
Double bib hood, for use without headband |
Each for $2,490.85
|
|
|||||
17-989-610
|
Bullard™
EVA20TICT |
Double bib hood, ratchet suspension, quick-attach |
Each for $2,495.96
|
|
|||||
Description
- Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
- Easy-to-read fuel gauge displays remaining battery capacity at a glance
- Battery status can be checked at any time during use
- Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
- Two distinct audible alarms notify wearers of low battery and low air flow
- Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
- Powered by a long-lasting 10,000-hour brushless motor
- Single filter design for easy use
- Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: (1) hood or mask; (2) breathing tube; (3) blower assembly; (4) purifying filter cartridge (sold separately). Check model system components carefully before ordering.
Compliance
NIOSH approved.
Specifications
7 to 8 hours | |
Foam | |
Acetate |