Bullard™ EVA PAPR System with Double Bib Tychem QC Hoods
Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) use a battery-powered fan to draw in ambient air through a filter cartridge then delivers filtered air to the user's double bib hood via a breathing tube.
Supplier:  Bullard™ EVART3T
 View more versions of this product
Catalog No. 17-989-615
Explore available promotions
Description
Description
  • Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
  • Easy-to-read fuel gauge displays remaining battery capacity at a glance
  • Battery status can be checked at any time during use
  • Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
  • Two distinct audible alarms notify wearers of low battery and low air flow
  • Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
  • Powered by a long-lasting 10,000-hour brushless motor
  • Single filter design for easy use
  • Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: (1) hood or mask; (2) breathing tube; (3) blower assembly; (4) purifying filter cartridge. Check model system components carefully before ordering.
Compliance
NIOSH approved.
Specifications
Specifications
7 to 8 hours
Double bib hood, for use without headband, quick-attach
Acetate
Foam
PAPR system, EVA blower, battery, charger, breathing tube
EVART3T