Caplugs™ (Evergreen Scientific) Histoware™ Prefilled 10% Formalin Vials
Contains half the nominal capacity of preservative
Supplier:  Caplugs™ (Evergreen Scientific) 3081103G8K
 View more versions of this product
Catalog No. 05-553-7
Explore available promotions
Description
Description
Wide-mouth openings permit easy loading of biopsy tissues and histological samples
  • Polyethylene screw-cap vials with wide-mouth openings
  • Permit easy loading of biopsy tissues and histological samples
  • Shatterproof, transparent
  • Polyethylene screwcaps have molded-in sealing rings for leakproof seals
05-553-1
7mL vials prefilled with 3.5mL 10% formalin plus one 100-position foam tray
05-553-7
20mL vials prefilled with 10mL 10% formalin plus two 20-position hard plastic processing trays
22-124-971
30mL vials prefilled with 15mL 10% formalin plus two 20-position hard plastic processing trays
Specifications
Specifications
20 mL, pre-filled 10 mL
Polyethylene
Screw Cap
Two 20-position hard plastic processing trays