Caplugs™ (Evergreen Scientific) Histoware™ Prefilled 10% Formalin Vials
Contains half the nominal capacity of preservative
$258.32 - $431.03
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Price | Quantity | |||||
05-553-7
|
Caplugs™ (Evergreen Scientific)
3081103G8K |
20 mL, pre-filled 10 mL | Two 20-position hard plastic processing trays |
Case of 100 Each for $258.32
|
|
|||||
05-553-1
|
Caplugs™ (Evergreen Scientific)
3081101G8T |
7 mL, pre-filled 3.5 mL | 100 count foam tray |
Case of 200 Each for $431.03
|
|
|||||
22-124-971
|
Caplugs™ (Evergreen Scientific)
3081107G8K |
30 mL | Includes two 20-position plastic trays |
Case of 100 Each for $289.09
|
|
|||||
Description
Wide-mouth openings permit easy loading of biopsy tissues and histological samples
- Polyethylene screw-cap vials with wide-mouth openings
- Permit easy loading of biopsy tissues and histological samples
- Shatterproof, transparent
- Polyethylene screwcaps have molded-in sealing rings for leakproof seals
05-553-1
7mL vials prefilled with 3.5mL 10% formalin plus one 100-position foam tray
05-553-7
20mL vials prefilled with 10mL 10% formalin plus two 20-position hard plastic processing trays
22-124-971
30mL vials prefilled with 15mL 10% formalin plus two 20-position hard plastic processing trays
Specifications
Polyethylene | |
Screw Cap |