missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Caplugs™ (Evergreen Scientific) Histoware™ Prefilled 10% Formalin Vials
Contains half the nominal capacity of preservative
Supplier: Caplugs™ (Evergreen Scientific) 3081107G8K
Description
Wide-mouth openings permit easy loading of biopsy tissues and histological samples
- Polyethylene screw-cap vials with wide-mouth openings
- Permit easy loading of biopsy tissues and histological samples
- Shatterproof, transparent
- Polyethylene screwcaps have molded-in sealing rings for leakproof seals
05-553-1
7mL vials prefilled with 3.5mL 10% formalin plus one 100-position foam tray
05-553-7
20mL vials prefilled with 10mL 10% formalin plus two 20-position hard plastic processing trays
22-124-971
30mL vials prefilled with 15mL 10% formalin plus two 20-position hard plastic processing trays
Specifications
30 mL | |
Polyethylene | |
Screw Cap | |
Includes two 20-position plastic trays |